2-(p-Tolyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

198870-5G
SIAL198870-5GEA 50.1 EUR
SIAL198870-5G
2-(p-Tolyl)pyridin, Sigma-Aldrich®
2-(p-Tolyl)pyridin
Formel: C₁₂H₁₁N
Molecular Weight: 169,22 g/mol
MDL: MFCD00006281
CAS-Nummer: 4467-06-5
EINECS: 224-734-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR