3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) 0.1 mol/l aqueous solution, Millipore®

Lieferant: Merck

Synonyme: CHAPS

19899-1ML-F
SIAL19899-1ML-FEA 64.9 EUR
SIAL19899-1ML-F
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS) 0.1 mol/l aqueous solution, Millipore®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel: C₃₂H₅₈N₂O₇S
Molecular Weight: 614.89 g/mol
Schmelzpunkt: 116
Lagertemperatur: Kühlschrank
MDL: MFCD00081083
CAS-Nummer: 75621-03-3
Merck Index: 13,02052

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR