Methyl-(4-methoxyphenyl)acetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-methoxyphenylacetate

M-Clarity™ Qualitätsstufe = MQ100

199206-25G
SIAL199206-25GEA 61.8 EUR
SIAL199206-25G
Methyl-(4-methoxyphenyl)acetat, Sigma-Aldrich®
Methyl-(4-methoxyphenyl)acetat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,23 g/mol
MDL: MFCD00008454
CAS-Nummer: 23786-14-3
EINECS: 245-886-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR