Chromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4H-Chromen-4-one

M-Clarity™ Qualitätsstufe = MQ100

199222-5G
SIAL199222-5GEA 77.7 EUR
SIAL199222-5G
Chromon, Sigma-Aldrich®
Chromon
Formel: C₉H₆O₂
Molecular Weight: 146,14 g/mol
MDL: MFCD00024064
CAS-Nummer: 491-38-3
EINECS: 207-737-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR