(S)-(+)-3-Amino-1-Boc-piperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-1-Boc-3-aminopiperidine, (S)-tert-Butyl 3-aminopiperidine-1-carboxylate, (S)-(+)-3-Amino-1-BOC-piperidine

19929-1G-F
SIAL19929-1G-FEA 83 EUR
SIAL19929-1G-F
(S)-(+)-3-Amino-1-Boc-piperidin, Sigma-Aldrich®
(S)-(+)-3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
MDL: MFCD03094718
CAS-Nummer: 625471-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR