12-Bromlaurinsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
200999-5G
SIAL200999-5GEA
328
EUR
SIAL200999-5G
12-Bromlaurinsäure, Sigma-Aldrich®
12-Bromlaurinsäure
Formel:
BrH₂C(CH₂)₁₀COOH Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002738 CAS-Nummer: 73367-80-3 EINECS: 277-401-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...