Arsen, pieces, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Arsenic solution
MQ100
202657-5G
SIAL202657-5GEA
87.5
EUR
SIAL202657-5G
Arsen, pieces, Sigma-Aldrich®
Arsenic
Formel:
As Molecular Weight: 74.92 g/mol Schmelzpunkt: 817 Dichte: 5.727 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085309 CAS-Nummer: 7440-38-2 EINECS: 231-148-6 UN: 1558 ADR: 6.1 II Merck Index: 12,00832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...