Cer(III)sulfat Octahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dicertrisulfat Octahydrat

M-Clarity™ Qualitätsstufe = MQ100

203009-50G
SIAL203009-50GEA 237 EUR
SIAL203009-50G
Cer(III)sulfat Octahydrat, Sigma-Aldrich®
Cer(III)sulfat Octahydrat
Formel: Ce₂O₁₂S₃·8H₂O
Molecular Weight: 712,54 g/mol
MDL: MFCD00149641
CAS-Nummer: 10450-59-6
EINECS: 236-644-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR