Erbiumtrichlorid Hexahydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
203211-20G
SIAL203211-20GEA
345
EUR
SIAL203211-20G
Erbiumtrichlorid Hexahydrat, Sigma-Aldrich®
Erbiumtrichlorid Hexahydrat
Formel:
Cl₃Er·6H₂O Molecular Weight: 381,71 g/mol Schmelzpunkt: 168 °C |
MDL:
MFCD00149693 CAS-Nummer: 10025-75-9 EINECS: 233-385-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...