Germanium, Pulver, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

203351-50G 203351-10G
SIAL203351-50GEA 1050 EUR
SIAL203351-50G SIAL203351-10G
Germanium, Pulver, Sigma-Aldrich®
Germanium
Formel: Ge
Lagertemperatur: Raumtemperatur
MDL: MFCD00085310
CAS-Nummer: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR