Diammoniumhexachloroiridat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ammoniumhexachloroiridat(IV)

203475-1G
SIAL203475-1GEA 320 EUR
SIAL203475-1G
Diammoniumhexachloroiridat, Sigma-Aldrich®
Diammoniumhexachloroiridat
Formel: H₈Cl₆IrN₂
Molecular Weight: 441,01 g/mol
Dichte: 2,85 g/cm³ (20 °C)
MDL: MFCD00010881
CAS-Nummer: 16940-92-4
EINECS: 241-007-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR