Mangan(II)nitrat Hydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Manganous nitrate hydrate
M-Clarity™ Qualitätsstufe = MQ100
203742-100G
203742-25G
SIAL203742-100GEA
500
EUR
SIAL203742-100G
SIAL203742-25G
Mangan(II)nitrat Hydrat, Sigma-Aldrich®
Mangan(II)nitrat Hydrat
Formel:
Mn(NO₃)₂·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149788 CAS-Nummer: 15710-66-4 EINECS: 233-828-8 UN: 2724 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...