Palladium(II)iodid, Sigma-Aldrich®

Lieferant: Merck

203963-2G
SIAL203963-2GEA 87.5 EUR
SIAL203963-2G
Palladium(II)iodid, Sigma-Aldrich®
Palladium(II)iodid
Formel: I₂Pd
Molecular Weight: 360,23 g/mol
Dichte: 6,003 g/cm³ (18 °C)
MDL: MFCD00011171
CAS-Nummer: 7790-38-7
EINECS: 232-203-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR