Samarium(III)chlorid Hexahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Samariumtrichlorid Hexahydrat

M-Clarity™ Qualitätsstufe = MQ200

204277-5G
SIAL204277-5GEA 71.5 EUR
SIAL204277-5G
Samarium(III)chlorid Hexahydrat, Sigma-Aldrich®
Samarium(III)chlorid Hexahydrat
Formel: Cl₃Sm·6H₂O
Molecular Weight: 364,81 g/mol
Dichte: 2,383 g/cm³ (25 °C)
MDL: MFCD00011235
CAS-Nummer: 13465-55-9
EINECS: 233-797-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR