Ytterbium(III)oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diytterbiumtrioxid, Ytterbiumoxid

M-Clarity™ Qualitätsstufe = MQ100

204889-10G
SIAL204889-10GEA 312 EUR
SIAL204889-10G
Ytterbium(III)oxid, Sigma-Aldrich®
Ytterbium(III)oxid
Formel: O₃Yb₂
Molecular Weight: 394,08 g/mol
Schmelzpunkt: 2346 °C
Dichte: 9,2 g/cm³ (20 °C)
MDL: MFCD00011290
CAS-Nummer: 1314-37-0
EINECS: 215-234-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR