4-Brombenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Bromophenyl)methanamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

205214-5G
SIAL205214-5GEA 107 EUR
SIAL205214-5G
4-Brombenzylamin Hydrochlorid, Sigma-Aldrich®
4-Brombenzylamin Hydrochlorid
Formel: C₇H₉BrClN
Molecular Weight: 222,52 g/mol
MDL: MFCD00012860
CAS-Nummer: 26177-44-6
EINECS: 247-503-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR