Blei(II)perchlorat Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
205311-250G
205311-50G
SIAL205311-250GEA
124
EUR
SIAL205311-250G
SIAL205311-50G
Blei(II)perchlorat Hydrat, Sigma-Aldrich®
Blei(II)perchlorat Hydrat
Formel:
Cl₂O₈Pb·H₂O |
CAS-Nummer:
207500-00-3 EINECS: 237-125-7 UN: 1470 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...