Dimethyl-2-aminoterephthalat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

M-Clarity™ Qualitätsstufe = MQ100

205370-50G
SIAL205370-50GEA 78.4 EUR
SIAL205370-50G
Dimethyl-2-aminoterephthalat, Sigma-Aldrich®
Dimethyl-2-aminoterephthalat
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.2 g/mol
MDL: MFCD00008427
CAS-Nummer: 5372-81-6
EINECS: 226-364-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR