(S)-(+)-3-tert-Butyl-2,5-piperazindion, Sigma-Aldrich®

Lieferant: Merck

20550-1G
SIAL20550-1GEA 385 EUR
SIAL20550-1G
(S)-(+)-3-tert-Butyl-2,5-piperazindion, Sigma-Aldrich®
(S)-(+)-3-tert-Butyl-2,5-piperazindion
Formel: C₈H₁₄N₂O₂
Molecular Weight: 170,21 g/mol
CAS-Nummer: 65050-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR