Palladium(II)nitrat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Palladiumdinitrat Hydrat

M-Clarity™ Qualitätsstufe = MQ100

205761-10G 205761-2G
SIAL205761-10GEA 1500 EUR
SIAL205761-10G SIAL205761-2G
Palladium(II)nitrat Hydrat, Sigma-Aldrich®
Palladium(II)nitrat Hydrat
Formel: N₂O₆Pd·xH₂O
Molecular Weight: 230.43 g/mol (anhy)
MDL: MFCD00149818
CAS-Nummer: 207596-32-5
EINECS: 233-265-8
UN: 1477
ADR: 5.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR