Palladium(II)oxid Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Palladiummonoxid Monohydrat

205842-2G
SIAL205842-2GEA 216 EUR
SIAL205842-2G
Palladium(II)oxid Monohydrat, Sigma-Aldrich®
Palladium(II)oxid Monohydrat
Formel: OPd·1H₂O
Molecular Weight: 140,43 g/mol
MDL: MFCD00011172
CAS-Nummer: 64109-12-2
EINECS: 215-218-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR