Butylsilan, Sigma-Aldrich®

Lieferant: Merck

20591-5ML-F
SIAL20591-5ML-FEA 164 EUR
SIAL20591-5ML-F
Butylsilan, Sigma-Aldrich®
Butylsilane
Formel: C₄H₁₂Si
Molecular Weight: 88,22 g/mol
MDL: MFCD00040079
CAS-Nummer: 1600-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR