Platin(II)iodid ≥98%, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Platindiiodid
M-Clarity™ Qualitätsstufe = MQ100
206059-1G
SIAL206059-1GEA
196
EUR
SIAL206059-1G
Platin(II)iodid ≥98%, Sigma-Aldrich®
Platin(II)iodid
Formel:
I₂Pt Molecular Weight: 448,89 g/mol Schmelzpunkt: 360 °C (Zers., Lit.) Dichte: 6,403 g/mL (25 °C) (Lit.) |
MDL:
MFCD00011183 CAS-Nummer: 7790-39-8 EINECS: 232-204-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...