Platin(II)bromid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
206121-1G
SIAL206121-1GEA
181
EUR
SIAL206121-1G
Platin(II)bromid, Sigma-Aldrich®
Platin(II)bromid
Formel:
Br₂Pt Molecular Weight: 354.89 g/mol Schmelzpunkt: 250 Dichte: 6.652 g/cm³ (25 °C) |
MDL:
MFCD00011180 CAS-Nummer: 13455-12-4 EINECS: 236-648-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...