1-Chlor-4-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Chlorphenyl)acetylen, 1-Ethinyl-4-chlorbenzol, (4-Chlorphenyl)ethin

206474-1G
SIAL206474-1GEA 34.4 EUR
SIAL206474-1G
1-Chlor-4-ethinylbenzol, Sigma-Aldrich®
1-Chlor-4-ethinylbenzol
Formel: C₈H₅Cl
Molecular Weight: 136,58 g/mol
MDL: MFCD00191917
CAS-Nummer: 873-73-4
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR