1-Brom-4-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethinyl-4-brombenzol, (4-Bromphenyl)acetylen, (4-Bromphenyl)ethin

M-Clarity™ Qualitätsstufe = MQ200

206512-1G
SIAL206512-1GEA 70.8 EUR
SIAL206512-1G
1-Brom-4-ethinylbenzol, Sigma-Aldrich®
1-Brom-4-ethinylbenzol
Formel: C₈H₅Br
Molecular Weight: 181,03 g/mol
MDL: MFCD00168822
CAS-Nummer: 766-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR