Vanadium(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
208272-25G
SIAL208272-25GEA
169
EUR
SIAL208272-25G
Vanadium(III)chlorid, Sigma-Aldrich®
Vanadium(III)chlorid
Formel:
Cl₃V Dichte: 3 g/cm³ (20 °C) |
MDL:
MFCD00011454 CAS-Nummer: 7718-98-1 EINECS: 231-744-6 UN: 2475 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...