Mangan(II)bromid Tetrahydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Mangandibromid Tetrahydrat
M-Clarity™ Qualitätsstufe = MQ100
208434-50G
SIAL208434-50GEA
70.9
EUR
SIAL208434-50G
Mangan(II)bromid Tetrahydrat, Sigma-Aldrich®
Mangan(II)bromid Tetrahydrat
Formel:
Br₂Mn·4H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149791 CAS-Nummer: 10031-20-6 EINECS: 236-591-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...