(2-Fluorphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid
M-Clarity™ Qualitätsstufe = MQ100
208949-10G
SIAL208949-10GEA
80.7
EUR
SIAL208949-10G
(2-Fluorphenyl)essigsäure, Sigma-Aldrich®
(2-Fluorphenyl)essigsäure
Formel:
(C₆H₄F)CH₂COOH Molecular Weight: 154,14 g/mol Siedepunkt: 263 °C (1013 hPa) Schmelzpunkt: 64 °C |
MDL:
MFCD00004315 CAS-Nummer: 451-82-1 EINECS: 207-196-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...