2,2-Dichlor-1-methylcyclopropancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

209767-25G
SIAL209767-25GEA 139 EUR
SIAL209767-25G
2,2-Dichlor-1-methylcyclopropancarbonsäure, Sigma-Aldrich®
2,2-Dichloro-1-methylcyclopropanecarboxylic acid
Formel: C₅H₆Cl₂O₂
Molecular Weight: 169,0059 g/mol
MDL: MFCD00001291
CAS-Nummer: 1447-14-9
EINECS: 215-901-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR