3-(1-Cyanethyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

209805-5G
SIAL209805-5GEA 82.6 EUR
SIAL209805-5G
3-(1-Cyanethyl)benzoesäure, Sigma-Aldrich®
3-(1-Cyanethyl)benzoesäure
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00002520
CAS-Nummer: 5537-71-3
EINECS: 226-897-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR