DL-α-(Methylaminomethyl)benzylalkohol, Sigma-Aldrich®

Lieferant: Merck

209848-10G
SIAL209848-10GEA 142 EUR
SIAL209848-10G
DL-α-(Methylaminomethyl)benzylalkohol, Sigma-Aldrich®
DL-α-(Methylaminomethyl)benzylalkohol
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00004506
CAS-Nummer: 6589-55-5
EINECS: 229-525-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR