5-Phenyl-o-anisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-4-phenylanisole, 2-Methoxy-5-phenylaniline, 5-Phenyl-o-anisidine, 3-Amino-4-methoxybiphenyl

M-Clarity™ Qualitätsstufe = MQ100

209880-1G
SIAL209880-1GEA 56 EUR
SIAL209880-1G
5-Phenyl-o-anisidin, Sigma-Aldrich®
5-Phenyl-o-anisidin
Formel: C₁₃H₁₃NO
Molecular Weight: 199,25 g/mol
MDL: MFCD00007790
CAS-Nummer: 39811-17-1
EINECS: 254-639-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR