Bortribromid 1 mol/l in Hexan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
211230-100ML
SIAL211230-100MLEA
156
EUR
SIAL211230-100ML
Bortribromid 1 mol/l in Hexan, Sigma-Aldrich®
Bortribromid
Formel:
BBr₃ Molecular Weight: 250.52 g/mol Siedepunkt: 91,2 Schmelzpunkt: -46 |
MDL:
MFCD00011312 CAS-Nummer: 10294-33-4 EINECS: 233-657-9 UN: 3384 ADR: 6.1 I Merck Index: 13,01337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...