3,5-Dinitro-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-methylbenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

211427-100G
SIAL211427-100GEA 48 EUR
SIAL211427-100G
3,5-Dinitro-o-toluylsäure, Sigma-Aldrich®
3,5-Dinitro-o-toluylsäure
Formel: C₈H₆N₂O₆
Molecular Weight: 226,15 g/mol
Schmelzpunkt: 203…207 °C
MDL: MFCD00007161
CAS-Nummer: 28169-46-2
EINECS: 248-880-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR