2,5-Dichlor-p-phenylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dichlor-1,4-phenylendiamin, 2,5-Dichlorbenzol-1,4-diamin, 1,4-Diamino-2,5-dichlorbenzol

M-Clarity™ Qualitätsstufe = MQ100

211494-25G
SIAL211494-25GEA 187 EUR
SIAL211494-25G
2,5-Dichlor-p-phenylendiamin, Sigma-Aldrich®
2,5-Dichlor-p-phenylendiamin
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,03 g/mol
MDL: MFCD00007902
CAS-Nummer: 20103-09-7
EINECS: 243-512-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR