Hydroxylapatit ≥90% (as Ca3(PO4)2, KT), purum p.a., Sigma-Aldrich®

Lieferant: Merck

21223-1KG
SIAL21223-1KGEA 66.8 EUR
SIAL21223-1KG
Hydroxylapatit ≥90% (as Ca3(PO4)2, KT), purum p.a., Sigma-Aldrich®
Hydroxylapatit
Formel: Ca₅HO₁₃P₃
MDL: MFCD00010904
CAS-Nummer: 1306-06-5
EINECS: 215-145-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR