2-Methyl-5-nitroanisol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Methoxy-2-methyl-5-nitrobenzene, Methyl 5-nitro-o-tolyl ether, 2-Methoxy-4-nitrotoluene

M-Clarity™ Qualitätsstufe = MQ200

213187-10G
SIAL213187-10GEA 0 EUR
SIAL213187-10G
2-Methyl-5-nitroanisol, Sigma-Aldrich®
2-Methyl-5-nitroanisol
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043912
CAS-Nummer: 13120-77-9
EINECS: 236-048-6
UN: 2446
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR