Antimon(III)ethoxid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

213314-5G
SIAL213314-5GEA 110 EUR
SIAL213314-5G
Antimon(III)ethoxid, Sigma-Aldrich®
Antimony (III) ethanolate
Formel: C₆H₁₅O₃Sb
MDL: MFCD00015123
CAS-Nummer: 10433-06-4
EINECS: 233-914-5
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR