1-Cyanpiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Cyanopiperidine, Pentamethylenecyanamide, Piperidine-1-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

213837-5G
SIAL213837-5GEA 118 EUR
SIAL213837-5G
1-Cyanpiperidin, Sigma-Aldrich®
1-Cyanpiperidin
Formel: C₆H₁₀N₂
MDL: MFCD00006477
CAS-Nummer: 1530-87-6
EINECS: 216-233-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR