4-Iod-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Iodopyrazole

M-Clarity™ Qualitätsstufe = MQ200

213993-10G
SIAL213993-10GEA 72.3 EUR
SIAL213993-10G
4-Iod-1H-pyrazol, Sigma-Aldrich®
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193,97 g/mol
MDL: MFCD00005244
CAS-Nummer: 3469-69-0
EINECS: 222-434-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR