2-Nitro-p-toluonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-2-nitrobenzonitril, 4-Methyl-2-nitrobenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ100

214299-1G
SIAL214299-1GEA 96.7 EUR
SIAL214299-1G
2-Nitro-p-toluonitril, Sigma-Aldrich®
2-Nitro-p-toluonitril
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00056107
CAS-Nummer: 26830-95-5
EINECS: 248-019-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR