3',5'-Dibenzyloxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-[3,5-Bis(benzyloxy)phenyl]ethan-1-one

M-Clarity™ Qualitätsstufe = MQ100

215341-50G
SIAL215341-50GEA 186 EUR
SIAL215341-50G
3',5'-Dibenzyloxyacetophenon, Sigma-Aldrich®
3',5'-Dibenzyloxyacetophenon
Formel: C₂₂H₂₀O₃
Molecular Weight: 332,40 g/mol
MDL: MFCD00004777
CAS-Nummer: 28924-21-2
EINECS: 249-315-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR