Methyl-2-amino-3,4,5-trimethoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-3,4,5-trimethoxybenzoesäuremethylester, Methyl-3,4,5-trimethoxyanthranilat

M-Clarity™ Qualitätsstufe = MQ100

215953-5G
SIAL215953-5GEA 94.8 EUR
SIAL215953-5G
Methyl-2-amino-3,4,5-trimethoxybenzoat, Sigma-Aldrich®
Methyl-2-amino-3,4,5-trimethoxybenzoat
Formel: C₁₁H₁₅NO₅
Molecular Weight: 241,24 g/mol
MDL: MFCD00008426
CAS-Nummer: 5035-82-5
EINECS: 225-728-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR