Bis(4-hydroxyphenyl)sulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-Thiodiphenol

M-Clarity™ Qualitätsstufe = MQ100

216178-100G
SIAL216178-100GEA 48.6 EUR
SIAL216178-100G
Bis(4-hydroxyphenyl)sulfid, Sigma-Aldrich®
Bis(4-hydroxyphenyl)sulfid
Formel: C₁₂H₁₀O₂S
Molecular Weight: 218,28 g/mol
MDL: MFCD00002349
CAS-Nummer: 2664-63-3
EINECS: 220-197-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR