3-Amino-5-methylcapronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-5-methylhexansäure

21625-1G
SIAL21625-1GEA 203 EUR
SIAL21625-1G
3-Amino-5-methylcapronsäure, Sigma-Aldrich®
3-Amino-5-methylcapronsäure
Formel: C₇H₁₅NO₂
Molecular Weight: 145,2 g/mol
Schmelzpunkt: 227 °C
MDL: MFCD01863281
CAS-Nummer: 3653-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR