Benzoesäure-[Ring-D5], Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,4,5,6-Pentadeuterobenzoesäure

M-Clarity™ Qualitätsstufe = MQ200

217158-1G
SIAL217158-1GEA 157 EUR
SIAL217158-1G
Benzoesäure-[Ring-D5], Sigma-Aldrich®
Benzoesäure-[Ring-D5]
Formel: C₆D₅COOH
Molecular Weight: 127,08 g/mol
Schmelzpunkt: 120…122 °C
CAS-Nummer: 1079-02-3
EINECS: 214-089-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR