Gadoliniumtrinitrat Hexahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Gadolinium (III) nitrate hexahydrate

217190-10G
SIAL217190-10GEA 85.1 EUR
SIAL217190-10G
Gadoliniumtrinitrat Hexahydrat, Sigma-Aldrich®
Gadoliniumtrinitrat Hexahydrat
Formel: GdN₃O₉·6H₂O
Molecular Weight: 451,36 g/mol
Schmelzpunkt: 91,5…92 °C
Dichte: 2,4 g/cm³ (20 °C)
MDL: MFCD00149728
CAS-Nummer: 19598-90-4
EINECS: 233-437-2
UN: 1477
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR