AmberChrom™ chloride form, 1X8 50 -100 Mesh, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

217417-500G 217417-100G 217417-2.5KG
SIAL217417-500GEA 426 EUR
SIAL217417-500G SIAL217417-100G SIAL217417-2.5KG
AmberChrom™ chloride form, 1X8 50 -100 Mesh, Supelco®
AmberChrom™ chloride form

AmberChrom™ 1X8 50-100 MESH (CL-) Anion is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.


Bitte beachten This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Formel: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Siedepunkt: 67 − 69 °C (11 mmHg)) (Lit.)
Schmelzpunkt: 5…8 °C (Lit.)
Dichte: 3,325 g/ml (25 °C) (Lit.)
CAS-Nummer: 69011-19-4

Bestellen

Spezifikation Testergebnisse

Form Liquid
Colour Almost colourless to yellow and almost colourless to green-yellow
Purity (GC) ≥98,5%
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR