2-Chlor-6-fluorphenylacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

218146-5G
SIAL218146-5GEA 23.6 EUR
SIAL218146-5G
2-Chlor-6-fluorphenylacetonitril, Sigma-Aldrich®
2-Chlor-6-fluorphenylacetonitril
Formel: C₈H₅ClFN
Molecular Weight: 169,59 g/mol
MDL: MFCD00001900
CAS-Nummer: 75279-55-9
EINECS: 278-171-8
UN: 3439
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR